4-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)morpholine 98%

Supplier: Ambeed
Warning

A910300-1G A910300-250MG A910300-5G
77159-830EA 40.17 USD
77159-830 77159-832 77159-834
4-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)morpholine 98%
4-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)morpholine

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₅S
MW: 353.24 g/mol
MDL Number: MFCD14702534
CAS Number: 1401222-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR