3,5-Difluoro-4-methoxybenzyl bromide 95%

Supplier: Ambeed
Danger

A909848-25G A909848-5G A909848-1G
77159-826EA 868 USD
77159-826 77159-828 77901-322
3,5-Difluoro-4-methoxybenzyl bromide 95%
3,5-Difluoro-4-methoxybenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MDL Number: MFCD04115916
CAS Number: 706786-42-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR