Fenoprofen calcium salt dihydrate 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Phenoxyphenyl)propionic Acid Calcium Salt, Calcium 2-(3-phenoxyphenyl)propanoate dihydrate

A905579-1G A905579-1MG A905579-250MG A905579-5G
77159-800EA 12 USD
77159-800 77159-802 77159-804 77159-806
Fenoprofen calcium salt dihydrate 98%
Fenoprofen calcium salt dihydrate

Precaución: for Research Use Only

Formula: C₃₀H₃₀CaO₈
MW: 558.63 g/mol
MDL Number: MFCD00242722
CAS Number: 71720-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR