L-Proline-p-nitroanilide trifluoroacetic acid salt 98%

Supplier: Ambeed
Warning

Synonyms: L-Proline p-nitroanilide trifluoroacetate salt, H-L-Pro-pNA Trifluoracetate

A905323-250MG A905323-5G A905323-1G
77159-796EA 41 USD
77159-796 77159-798 77614-624
L-Proline-p-nitroanilide trifluoroacetic acid salt 98%
L-Proline-p-nitroanilide trifluoroacetic acid salt

Precaución: for Research Use Only

Formula: C₁₃H₁₄F₃N₃O₅
MW: 349.266 g/mol
MDL Number: MFCD00069671
CAS Number: 108321-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR