Dibenzo[b,d]thiophene-2,8-diylbis(diphenylphosphine oxide) 95%
Supplier: Ambeed
|
Danger
|
A905068-100MG
A905068-1G
A905068-250MG
A905068-5G
77159-788EA
41.2
USD
77159-788
77159-790
77159-792
77159-794
Dibenzo[b,d]thiophene-2,8-diylbis(diphenylphosphine oxide) 95%
Dibenzo[b,d]thiophene-2,8-diylbis(diphenylphosphine oxide)
Precaución: for Research Use Only
Formula:
C₃₆H₂₆O₂P₂S MW: 584.60 g/mol |
MDL Number:
MFCD27978274 CAS Number: 1019842-99-9 UN: 2811 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...