1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulphonate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Butyl-2,3-dimethylimidazolium triflate, 1-Butyl-2,3-dimethylimidazolium Trifluoromethanesulphonate
A902820-100G
A902820-25G
A902820-5G
77159-782EA
223
USD
77159-782
77159-784
77159-786
1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulphonate 98%
1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulphonate
Precaución: for Research Use Only
Formula:
C₁₀H₁₇F₃N₂O₃S MW: 302.32 g/mol |
MDL Number:
MFCD03427624 CAS Number: 765910-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...