(1S,3R,4R)-Ethyl 3-((tert-butoxycarbonyl)amino)-4-hydroxycyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

A899313-100G A899313-25G A899313-5G
77159-772EA 194 USD
77159-772 77159-774 77159-776
(1S,3R,4R)-Ethyl 3-((tert-butoxycarbonyl)amino)-4-hydroxycyclohexanecarboxylate 97%
(1S,3R,4R)-Ethyl 3-((tert-butoxycarbonyl)amino)-4-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₅
MDL Number: MFCD23106245
CAS Number: 365997-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR