Disodium hydrogen phosphate heptahydrate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate

A898590-100G A898590-25G A898590-500G A898590-5G A898590-1000G
77159-764EA 12 USD
77159-764 77159-766 77159-768 77159-770 77595-444
Disodium hydrogen phosphate heptahydrate 99%
Disodium hydrogen phosphate heptahydrate

Precaución: for Research Use Only

Formula: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Melting Pt: 48
MDL Number: MFCD00149180
CAS Number: 7782-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR