(E)-3-(4-((E)-2-(2-Chloro-4-fluorophenyl)-1-(1H-indazol-5-yl)but-1-en-1-yl)phenyl)acrylic acid 98+%

Supplier: Ambeed
Warning

Synonyms: Brilanestrant

A897745-10MG A897745-1MG A897745-25MG A897745-5MG A897745-50MG
77159-756EA 91 USD
77159-756 77159-758 77159-760 77159-762 77900-529
(E)-3-(4-((E)-2-(2-Chloro-4-fluorophenyl)-1-(1H-indazol-5-yl)but-1-en-1-yl)phenyl)acrylic acid 98+%
(E)-3-(4-((E)-2-(2-Chloro-4-fluorophenyl)-1-(1H-indazol-5-yl)but-1-en-1-yl)phenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₀ClFN₂O₂
MDL Number: MFCD28902190
CAS Number: 1365888-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR