2-(Boc-amino)-4-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-[(tert-Butoxycarbonyl)amino]-4-methylpyridine, 2-(Boc-amino)-4-methylpyridine, (4-Methylpyridin-2-yl)carbamic acid tert-Butyl ester, tert-Butyl (4-Methylpyridin-2-yl)carbamate

A894914-5G A894914-25G A894914-1G
77159-742EA 127.72 USD
77159-742 77614-602 77614-600
2-(Boc-amino)-4-methylpyridine 95%
2-(Boc-amino)-4-methylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O₂
MW: 208.261 g/mol
MDL Number: MFCD07776933
CAS Number: 90101-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR