3-Fluoro-4-nitrobenzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 3-Fluoro-4-nitrobenzene-1-sulphonyl chloride

A889337-100MG A889337-10G A889337-1G A889337-250MG A889337-25G
77159-728EA 15.45 USD
77159-728 77159-730 77159-732 77159-734 77686-928
3-Fluoro-4-nitrobenzenesulfonyl chloride 95%
3-Fluoro-4-nitrobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₄S
MW: 239.61 g/mol
MDL Number: MFCD03412240
CAS Number: 86156-93-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR