2-Chloro-1-iodo-4-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-iodonitrobenzene

A889259-100G A889259-10G A889259-25G A889259-5G
77159-720EA 330.63 USD
77159-720 77159-722 77159-724 77159-726
2-Chloro-1-iodo-4-nitrobenzene 98%
2-Chloro-1-iodo-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClINO₂
MW: 283.45 g/mol
MDL Number: MFCD00024589
CAS Number: 41252-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR