5-Bromo-1,3-dimethyl-2-pyridone 95%
Supplier: Ambeed
|
Warning
|
A886137-100MG
A886137-1G
A886137-250MG
A886137-5G
77159-702EA
8
USD
77159-702
77159-704
77159-706
77159-708
5-Bromo-1,3-dimethyl-2-pyridone 95%
5-Bromo-1,3-dimethyl-2-pyridone
Precaución: for Research Use Only
Formula:
C₇H₈BrNO |
MDL Number:
MFCD16658881 CAS Number: 51417-13-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...