(Ac-Cys-OH)₂ 97%

Supplier: Ambeed
Warning

A884143-100MG A884143-1G A884143-250MG A884143-5G
77159-672EA 45 USD
77159-672 77159-674 77159-676 77159-678
(Ac-Cys-OH)₂ 97%
(Ac-Cys-OH)₂

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₆S₂
MDL Number: MFCD00152053
CAS Number: 5545-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR