Methyl-3-oxo-1-methylcyclobutanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-3-oxocyclobutanecarboxylate

A883450-250MG A883450-5G
77159-668EA 37.08 USD
77159-668 77159-670
Methyl-3-oxo-1-methylcyclobutanecarboxylate 98%
Methyl-3-oxo-1-methylcyclobutanecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MW: 142.16 g/mol
MDL Number: MFCD23105986
CAS Number: 1408075-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR