6-Methyl-1H-indole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A883130-100MG A883130-250MG
77159-664EA 176.13 USD
77159-664 77159-666
6-Methyl-1H-indole-4-carboxylic acid 95%
6-Methyl-1H-indole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.18 g/mol
MDL Number: MFCD29037436
CAS Number: 1546171-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR