4-((4-(1H-Tetrazol-1-yl)phenoxy)methyl)-2-(1-(5-ethylpyrimidin-2-yl)piperidin-4-yl)thiazole 98%

Supplier: Ambeed
Danger

Synonyms: MBX-2982

A882623-10MG A882623-1MG A882623-25MG A882623-5MG
77159-656EA 90.64 USD
77159-656 77159-658 77159-660 77159-662
4-((4-(1H-Tetrazol-1-yl)phenoxy)methyl)-2-(1-(5-ethylpyrimidin-2-yl)piperidin-4-yl)thiazole 98%
4-((4-(1H-Tetrazol-1-yl)phenoxy)methyl)-2-(1-(5-ethylpyrimidin-2-yl)piperidin-4-yl)thiazole

Precaución: for Research Use Only

Formula: C₂₂H₂₄N₈OS
MW: 448.54 g/mol
MDL Number: MFCD22628771
CAS Number: 1037792-44-1
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR