3-[4-(Methylsulfonyl)phenyl]-5-(1-boc-4-piperidyl)-1,2,4-oxadiazole 97%

Supplier: Ambeed
Warning

A881508-100MG A881508-1G A881508-250MG
77159-642EA 9 USD
77159-642 77159-644 77159-646
3-[4-(Methylsulfonyl)phenyl]-5-(1-boc-4-piperidyl)-1,2,4-oxadiazole 97%
3-[4-(Methylsulfonyl)phenyl]-5-(1-boc-4-piperidyl)-1,2,4-oxadiazole

Precaución: for Research Use Only

Formula: C₁₉H₂₅N₃O₅S
MDL Number: MFCD18651706
CAS Number: 1272756-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR