UNC0646 98%

Supplier: Ambeed
Warning

Synonyms: N-​(1-​cyclohexyl-​4-​piperidinyl)-​2-​[hexahydro-​4-​(1-​methylethyl)-​1H-​1,​4-​diazepin-​1-​yl]-​6-​methoxy-​7-​[3-​(1-​piperidinyl)propoxy]-​4-​quinazolinamine, N-(1-Cyclohexylpiperidin-4-yl)-2-(4-isopropyl-1,4-diazepan-1-yl)-6-methoxy-7-(3-(piperidin-1-yl)propoxy)quinazolin-4-amine

A880677-10MG A880677-1MG A880677-25MG A880677-5MG A880677-50MG
77159-634EA 57 USD
77159-634 77159-636 77159-638 77159-640 77901-828
UNC0646 98%
UNC0646

Precaución: for Research Use Only

Formula: C₃₆H₅₉N₇O₂
MW: 621.90 g/mol
MDL Number: MFCD22378486
CAS Number: 1320288-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR