5-Bromo-2-(difluoromethyl)-1,3-difluorobenzene 95%

Supplier: Ambeed
Warning

A880455-100MG A880455-250MG A880455-5G A880455-1G
77159-628EA 26.78 USD
77159-628 77159-630 77159-632 77686-920
5-Bromo-2-(difluoromethyl)-1,3-difluorobenzene 95%
5-Bromo-2-(difluoromethyl)-1,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₇H₃BrF₄
MDL Number: MFCD16140200
CAS Number: 1221272-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR