3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A876136-10G A876136-25G A876136-5G
77159-622EA 23.69 USD
77159-622 77159-624 77159-626
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine 98%
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₄N
MW: 199.53 g/mol
MDL Number: MFCD00191918
CAS Number: 72537-17-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR