Ethyl 8-hydroxyquinoline-5-carboxylate 95%

Supplier: Ambeed
Warning

A874502-100MG A874502-250MG
77159-608EA 160 USD
77159-608 77159-610
Ethyl 8-hydroxyquinoline-5-carboxylate 95%
Ethyl 8-hydroxyquinoline-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD18416650
CAS Number: 104293-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR