6-Methyl-5-nitroisoquinoline 95%

Supplier: Ambeed
Warning

A873573-250MG A873573-5G A873573-1G
77159-604EA 15 USD
77159-604 77159-606 77686-912
6-Methyl-5-nitroisoquinoline 95%
6-Methyl-5-nitroisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD11848649
CAS Number: 188121-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR