Fmoc-Thz-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-L-thiazolidine-4-carboxylic acid, Fmoc-L-thioproline, (R)-3-(((9H-Fluoren-9-yl)methoxy)carbonyl)thiazolidine-4-carboxylic acid, (4R)-3-[(9H-Fluoren-9-ylmethoxy)carbonyl]-1,3-thiazolane-4-carboxylic acid

A872504-5G A872504-100G A872504-1G A872504-25G
77159-596EA 19 USD
77159-596 77686-911 77901-248 77901-595
Fmoc-Thz-OH 98%
Fmoc-Thz-OH

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₄S
MW: 355.408 g/mol
MDL Number: MFCD00235910
CAS Number: 133054-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR