Methyl 3-nitro-4-(trifluoromethyl)benzoate 95%

Supplier: Ambeed
Warning

A870696-10G A870696-1G A870696-25G A870696-5G
77159-578EA 44 USD
77159-578 77159-580 77159-582 77159-584
Methyl 3-nitro-4-(trifluoromethyl)benzoate 95%
Methyl 3-nitro-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆F₃NO₄
MDL Number: MFCD09264510
CAS Number: 126541-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR