(4-Phenyl-1,3-thiazol-2-yl)methanamine 95%

Supplier: Ambeed
Danger

Synonyms: (4-Phenylthiazol-2-yl)methanamine

A866167-100MG A866167-250MG
77159-564EA 37.08 USD
77159-564 77159-566
(4-Phenyl-1,3-thiazol-2-yl)methanamine 95%
(4-Phenyl-1,3-thiazol-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S
MW: 190.265 g/mol
MDL Number: MFCD05221867
CAS Number: 90916-45-3
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR