1-Naphthyltrifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

A864921-1G A864921-25G A864921-5G
77159-558EA 34 USD
77159-558 77159-560 77159-562
1-Naphthyltrifluoromethanesulfonate 95%
1-Naphthyltrifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₇F₃O₃S
MDL Number: MFCD00192340
CAS Number: 99747-74-7
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR