4-(Trifluoromethyl)-DL-phenylalanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-DL-Phe(4-CF₃)-OH
A863277-1G
A863277-250MG
77159-556EA
86
USD
77159-556
77686-903
4-(Trifluoromethyl)-DL-phenylalanine 98%
4-(Trifluoromethyl)-DL-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₀H₁₀F₃NO₂ MW: 233.19 g/mol |
MDL Number:
MFCD00070813 CAS Number: 14091-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...