2-Fluoro-5-nitrobenzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

A858474-1G A858474-250MG
77159-536EA 153.47 USD
77159-536 77159-538
2-Fluoro-5-nitrobenzenesulfonyl chloride 95%
2-Fluoro-5-nitrobenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₄S
MW: 239.609 g/mol
MDL Number: MFCD08443561
CAS Number: 713-21-3
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR