4-(2-Fluorophenoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

A849071-100MG A849071-1G A849071-250MG
77159-498EA 35.02 USD
77159-498 77159-500 77159-502
4-(2-Fluorophenoxy)phenylboronic acid 98%
4-(2-Fluorophenoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BFO₃
MW: 232.02 g/mol
MDL Number: MFCD18311840
CAS Number: 1402238-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR