3-Bromo-4-methoxy-1H-pyrrolo[2,3-b]pyridine 98%
Supplier: Ambeed
|
Warning
|
A844874-100MG
A844874-1G
A844874-250MG
A844874-5G
77159-490EA
19
USD
77159-490
77159-492
77159-494
77159-496
3-Bromo-4-methoxy-1H-pyrrolo[2,3-b]pyridine 98%
3-Bromo-4-methoxy-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₈H₇BrN₂O |
MDL Number:
MFCD10699217 CAS Number: 1053655-78-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...