N-(2,3,4-Trifluorophenyl)acetamide 96%

Supplier: Ambeed
Warning

A844755-1G A844755-250MG A844755-5G A844755-25G
77159-484EA 37 USD
77159-484 77159-486 77159-488 77893-650
N-(2,3,4-Trifluorophenyl)acetamide 96%
N-(2,3,4-Trifluorophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₆F₃NO
MDL Number: MFCD01935687
CAS Number: 365-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR