TUG-891 95%

Supplier: Ambeed
Warning

Synonyms: 4-[(4-Fluoro-4′-methyl[1,1′-biphenyl]-2-yl)methoxy]-benzenepropanoic acid, 3-(4-((4-Fluoro-4'-methyl-[1,1'-biphenyl]-2-yl)methoxy)phenyl)propanoic acid, 3-(4-((4-Fluoro-4-methyl-[1,1-biphenyl]-2-yl)methoxy)phenyl)-propanoic acid

A844329-10MG A844329-1MG A844329-25MG A844329-50MG A844329-5MG
77159-474EA 37.08 USD
77159-474 77159-476 77159-478 77159-480 77159-482
TUG-891 95%
TUG-891

Precaución: for Research Use Only

Formula: C₂₃H₂₁FO₃
MW: 364.41 g/mol
MDL Number: MFCD22421654
CAS Number: 1374516-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR