5-sulfamoyl-furan-3-carboxylic acid ethyl ester 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-sulphamoylfuran-3-carboxylate, Ethyl 5-sulphamoyl-3-furoate

A843896-100MG A843896-250MG
77159-470EA 96.82 USD
77159-470 77159-472
5-sulfamoyl-furan-3-carboxylic acid ethyl ester 95%
5-sulfamoyl-furan-3-carboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₇H₉NO₅S
MW: 219.215 g/mol
MDL Number: MFCD24386139
CAS Number: 256373-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR