8-Aminoquinoline-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A842237-100MG A842237-250MG A842237-1G A842237-5G
77159-466EA 124.63 USD
77159-466 77159-468 77552-572 77552-574
8-Aminoquinoline-4-carboxylic acid 98%
8-Aminoquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
MDL Number: MFCD08690743
CAS Number: 121689-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR