Boc-Lys(Ac)-AMC 95%

Supplier: Ambeed
Warning

A840516-100MG A840516-250MG A840516-1G
77159-458EA 245 USD
77159-458 77686-871 77901-029
Boc-Lys(Ac)-AMC 95%
Boc-Lys(Ac)-AMC

Precaución: for Research Use Only

Formula: C₂₃H₃₁N₃O₆
MDL Number: MFCD02683946
CAS Number: 233691-67-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR