cis-4-Aminotetrahydropyran-3-ol hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (3R,4R)-4-Aminotetrahydro-2H-pyran-3-ol hydrochloride

A837187-1G A837187-250MG A837187-5G
77159-446EA 203.94 USD
77159-446 77159-448 77159-450
cis-4-Aminotetrahydropyran-3-ol hydrochloride 95%
cis-4-Aminotetrahydropyran-3-ol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO₂
MW: 153.61 g/mol
MDL Number: MFCD28099084
CAS Number: 1523530-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR