4-Chloro-6-nitroquinoline 98%

Supplier: Ambeed
Warning

A836784-1G A836784-250MG
77159-438EA 141.11 USD
77159-438 77614-500
4-Chloro-6-nitroquinoline 98%
4-Chloro-6-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
MW: 208.601 g/mol
MDL Number: MFCD00498990
CAS Number: 13675-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR