4-Methyl-2-(piperidin-1-yl)quinoline 98%

Supplier: Ambeed
Warning

A835423-10MG A835423-1MG A835423-25MG A835423-50MG A835423-5MG
77159-426EA 20 USD
77159-426 77159-428 77159-430 77159-432 77159-434
4-Methyl-2-(piperidin-1-yl)quinoline 98%
4-Methyl-2-(piperidin-1-yl)quinoline

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂
MDL Number: MFCD00817920
CAS Number: 5465-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR