2,6-Dichloro-3-bromobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A833553-100MG
A833553-1G
A833553-250MG
A833553-5G
77159-414EA
9
USD
77159-414
77159-416
77159-418
77893-625
2,6-Dichloro-3-bromobenzoic acid 98%
2,6-Dichloro-3-bromobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃BrCl₂O₂ |
MDL Number:
MFCD18432783 CAS Number: 80257-12-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...