Fmoc-3-amino-3-(4-bromophenyl)propionic acid 95%
Supplier: Ambeed
|
Warning
|
A832096-1G
A832096-5G
A832096-250MG
77159-402EA
80
USD
77159-402
77159-404
77901-083
Fmoc-3-amino-3-(4-bromophenyl)propionic acid 95%
Fmoc-3-amino-3-(4-bromophenyl)propionic acid
Precaución: for Research Use Only
Formula:
C₂₄H₂₀BrNO₄ |
MDL Number:
MFCD01863199 CAS Number: 269078-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...