tert-Butyl 2-formyl-6-azaspiro[3.4]octane-6-carboxylate 95%

Supplier: Ambeed
Warning

A831429-100MG A831429-1G A831429-250MG
77159-396EA 45 USD
77159-396 77159-398 77159-400
tert-Butyl 2-formyl-6-azaspiro[3.4]octane-6-carboxylate 95%
tert-Butyl 2-formyl-6-azaspiro[3.4]octane-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₃
MDL Number: MFCD24857296
CAS Number: 203662-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR