1-(5-Chloro-2-pyridinyl)piperazine 98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Chloro-2-pyridyl)piperazine

A830131-1G A830131-250MG A830131-5G
77159-388EA 53.56 USD
77159-388 77159-390 77159-392
1-(5-Chloro-2-pyridinyl)piperazine 98%
1-(5-Chloro-2-pyridinyl)piperazine

Precaución: for Research Use Only

Formula: C₉H₁₂ClN₃
MW: 197.67 g/mol
MDL Number: MFCD04115049
CAS Number: 87394-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR