3-Acetyl-4-chloro-7-azaindole 97%

Supplier: Ambeed
Warning

A827643-1G A827643-250MG
77159-384EA 125.66 USD
77159-384 77159-386
3-Acetyl-4-chloro-7-azaindole 97%
3-Acetyl-4-chloro-7-azaindole

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MW: 194.618 g/mol
MDL Number: MFCD12828645
CAS Number: 1011711-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR