(3-Bromo-5-formylphenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-formylphenylboronic acid

A827564-100MG A827564-1G A827564-250MG
77159-378EA 15 USD
77159-378 77159-380 77159-382
(3-Bromo-5-formylphenyl)boronic acid 95%
(3-Bromo-5-formylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₆BBrO₃
MW: 228.83 g/mol
MDL Number: MFCD10696657
CAS Number: 157866-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR