cis-4-Methoxycyclohexanamine hydrochloride 97%

Supplier: Ambeed
Warning

A827280-1G A827280-250MG
77159-374EA 151 USD
77159-374 77159-376
cis-4-Methoxycyclohexanamine hydrochloride 97%
cis-4-Methoxycyclohexanamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO
MDL Number: MFCD18207780
CAS Number: 61367-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR