4-Bromo-5-nitro-1H-indazole 97%

Supplier: Ambeed
Warning

A817913-100MG A817913-1G A817913-250MG
77159-330EA 47.38 USD
77159-330 77159-332 77159-334
4-Bromo-5-nitro-1H-indazole 97%
4-Bromo-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.02 g/mol
MDL Number: MFCD12963526
CAS Number: 1190315-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR