(R)-2-Methylpiperidin-4-one hydrochloride 98%

Supplier: Ambeed
Warning

A816217-100MG A816217-1G A816217-250MG A816217-5G
77159-324EA 9 USD
77159-324 77159-326 77159-328 77893-602
(R)-2-Methylpiperidin-4-one hydrochloride 98%
(R)-2-Methylpiperidin-4-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO
MDL Number: MFCD22682261
CAS Number: 1434126-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR