3-Amino-2-chlorobenzoic acid 98%

Supplier: Ambeed
Warning

A815228-10G A815228-1G A815228-250MG A815228-25G A815228-5G
77159-312EA 317.24 USD
77159-312 77159-314 77159-316 77159-318 77159-320
3-Amino-2-chlorobenzoic acid 98%
3-Amino-2-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C7H6ClNO2
MW: 171.5810 g/mol
MDL Number: MFCD00191726
CAS Number: 108679-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR