Thymol blue sodium salt (red) 1.2 to 2.8 (yellow), (yellow) 8.0 to 9.6 (blue)
Supplier: Ambeed
|
Warning
|
A814556-25G
A814556-5G
A814556-100G
77159-308EA
45
USD
77159-308
77159-310
77901-352
Thymol blue sodium salt (red) 1.2 to 2.8 (yellow), (yellow) 8.0 to 9.6 (blue)
Thymol blue sodium salt
Precaución: for Research Use Only
Formula:
C₂₇H₂₉NaO₅S |
MDL Number:
MFCD00151093 CAS Number: 62625-21-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...